Order Entry
Germany
ContactUsLinkComponent
Chloro(1,5-cyclooctadiene)rhodium(I) dimer
Chloro(1,5-cyclooctadiene)rhodium(I) dimer
Catalog # COBBST-7037-1G
Supplier:  COMBI-BLOCKS
CAS Number:  
Chloro(1,5-cyclooctadiene)rhodium(I) dimer
Catalog # COBBST-7037-1G
Supplier:  COMBI-BLOCKS
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :493.08 g/mol
EINECS:235-157-6
CAS Number:12092-47-6
Formula:C₁₆H₂₄Cl₂Rh₂
MDL Number:MFCD00012415