2-Amino-5-methoxypyrazine
Catalog # COBBQA-8419-1G
Supplier: COMBI-BLOCKS
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:125.13 g/mol
CAS Number:54013-07-9
Formula:C₅H₇N₃O
MDL Number:MFCD09909647