Order Entry
Germany
ContactUsLinkComponent
Mercury standard solution, 10 mg/L Hg in dil. nitric acid (from Hg)
Mercury standard solution, 10 mg/L Hg in dil. nitric acid (from Hg)
Catalog # PERCN9300253
Supplier:  PerkinElmer
CAS Number:  
Mercury standard solution, 10 mg/L Hg in dil. nitric acid (from Hg)
Catalog # PERCN9300253
Supplier:  PerkinElmer
CAS Number:  

Specifications

  • Concentration:
    10 mg/L
  • Matrix:
    Nitric acid dil.
  • Pk:
    1
  • Element:
    Mercury
  • Symbol:
    Hg

Specifications

About this item