1,3-Dichloro-2-fluorobenzene
Catalog # COBBOR-2043-100G
Supplier: COMBI-BLOCKS
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:164.99 g/mol
Boiling Pt:168 °C (1013 hPa)
Melting Pt:39 °C
UN:1325
ADR:4.1,II
:GHS07
CAS Number:2268-05-5
Formula:C₆H₃Cl₂F
MDL Number:MFCD00010307