

Chloroform-[D₁] (99.8% D) + 0.03% (v/v) TMS, AcroSeal® for NMR spectroscopy
Catalog # 426961000.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Water:≤0.02 %
- D-Enrichment:≥99.75 %
- TMS:0.028 to 0.032 v/v%
Specification Test Results
About this item
:
:120.38 g/mol
Boiling Pt:60.8 °C (1013 hPa)
Melting Pt:–64 °C
Density:1.5 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:212-742-4
UN:1888
ADR:6.1,III
:GHS06
:GHS08
CAS Number:865-49-6
Formula:CDCl₃
MDL Number:MFCD00000827