Order Entry
Germany
ContactUsLinkComponent
 
Trifluoroacetic anhydride
Catalog # ABCRAB212439.0500
Supplier:  abcr
CAS Number:  
Trifluoroacetic anhydride
Catalog # ABCRAB212439.0500
Supplier:  abcr
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications

About this item

 :
 :210.03 g/mol
Boiling Pt:39.4 °C (1013 hPa)
Melting Pt:–65 °C
Density:1.487 g/cm³ (20 °C)
EINECS:206-982-9
UN:3265
ADR:8,I
 :GHS05
 :GHS07
CAS Number:407-25-0
Formula:C₄F₆O₃
MDL Number:MFCD00000416