Order Entry
Germany
ContactUsLinkComponent
D-(+)-Glucose 10% in aqueous solution, Sigma-Aldrich®
D-(+)-Glucose 10% in aqueous solution, Sigma-Aldrich®
Catalog # SIALG8644-100ML
Supplier:  Merck
CAS Number:  
undefined
D-(+)-Glucose 10% in aqueous solution, Sigma-Aldrich®
Catalog # SIALG8644-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ300
 :180.16 g/mol
Storage Temperature:Ambient
EINECS:200-075-1
CAS Number:50-99-7
 :492-62-6
Formula:C₆H₁₂O₆
MDL Number:MFCD00148912