Order Entry
Germany
Orders LinkContactUsLinkComponent
2-(Ammoniooxy)propane chloride

2-(Ammoniooxy)propane chloride

Marketsource
Supplier:   BIONET RESEARCH
2-(Ammoniooxy)propane chloride
Supplier:  BIONET RESEARCH
Marketsource
Certificates
2-(Ammoniooxy)propane chloride

4 Options Available Below

Product Details & Documents
Synonyms:  O-propan-2-ylhydroxylamine hydrochloride, O-Isopropylhydroxylamine hydrochloride
  • CAS Number:4490-81-7
  • Formula:C₃H₁₀ClNO
  • MDL Number:MFCD00231457
  • Molecular Weight:111.5706 g/mol
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    500 mg
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.