Order Entry
Germany
Orders LinkContactUsLinkComponent
4-Hydroxy-2,6-dimethoxybenzaldehyde

4-Hydroxy-2,6-dimethoxybenzaldehyde

Glass bottle for solids, 1 g
Catalog # MOLE18893113-1G
Marketsource
Supplier:   Molekula
4-Hydroxy-2,6-dimethoxybenzaldehyde
Catalog # MOLE18893113-1G
Supplier:  Molekula
Marketsource
CAS Number:  22080-96-2
4-Hydroxy-2,6-dimethoxybenzaldehyde

Glass bottle for solids, 1 g

CAS Number:  22080-96-2
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  2,6-Dimethoxy-4-hydroxybenzaldehyde
  • CAS Number:22080-96-2
  • Formula:C₉H₁₀O₄
  • MDL Number:MFCD01407645
  • Molecular Weight:182.1733 g/mol
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.