Order Entry
Germany
ContactUsLinkComponent
Pesticide reference standard, 1,2,3-Trichlorobenzene 1,000 µg/ml in cyclohexane
Pesticide reference standard, 1,2,3-Trichlorobenzene 1,000 µg/ml in cyclohexane
  PROONE5708
 :  
Pesticide reference standard, 1,2,3-Trichlorobenzene 1,000 µg/ml in cyclohexane
  PROONE5708
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1,5 mL
  • CAS No.:
    87-61-6
  • Description:
    1,2,3-Trichlorobenzene 1,000 µg/ml in cyclohexane
  • Formula:
    C₆H₃Cl₃
  • Description:
    1,2,3-Trichlorobenzene