(Benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (BOP)
Catalog # UPST20-173
Supplier: UPSTATE TECHNOLOGY
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:442.28 g/mol
EINECS:260-279-1
CAS Number:56602-33-6
Formula:C₁₂H₂₂F₆N₆OP₂
MDL Number:MFCD00011948