Auftragserfassung
Germany
ContactUsLinkComponent
 
 
 
(Benzotriazol-1-yloxy)tris(dimethylamino)phosphoniumhexafluorophosphat (BOP) 98+%
 
undefined
 
 
(Benzotriazol-1-yloxy)tris(dimethylamino)phosphoniumhexafluorophosphat (BOP) 98+%

 

  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    136 to 140 °C
  • HPLC:
    ≥98.0 %

 

 

Frequently Bought Together