Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
L(-)-Bornyl acetate
L(-)-Bornyl acetate, Supelco®
L(-)-Bornyl acetate, Supelco®
L(-)-Bornyl acetate, Supelco®
Supplier:
Merck
Certificates
Expand image
L(-)-Bornyl acetate, Supelco®
Supplier:
Merck
Certificates
About this item
CAS Number:
5655-61-8
:
Formula:
C₁₂H₂₀O₂
MDL Number:
MFCD00867808
:
196.29 g/mol
:
M-Clarity™ quality level = MQ100
EINECS:
227-101-4
Related Information
Documentation
Certificates
Search by Lot Number
Documentation