L(-)-Bornyl acetate 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5655-61-8
:
Formula:C₁₂H₂₀O₂
MDL Number:MFCD00867808
:196.29 g/mol
Boiling Pt:223…224 °C (760 mmHg)
Density:0.9875 g/cm³
Flash Pt:84 °C
Storage Temperature:Ambient
EINECS:227-101-4
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4620 to 1.4650 (20 °C, 589 nm)
- GC:≥94 %
- Specific gravity:(15/15 °C) 0.986 to 0.99
- Specific optical rotation:(20 °C, 589 nm) (neat) < -38°