1-(2-(Ammoniooxy)ethoxy)-3-(trifluoromethyl)benzene chloride
Catalog # FLUO010008-1G
Supplier: FLUOROCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
CAS Number:113211-36-2
Formula:C₉H₁₁ClF₃NO₂
MDL Number:MFCD01114594