Order Entry
Germany
ContactUsLinkComponent
Ammonium chloride-[¹⁵N], Overnight Express™, Sigma-Aldrich®
Ammonium chloride-[¹⁵N], Overnight Express™, Sigma-Aldrich®
Catalog # 71759-4
CAS Number:  
Ammonium chloride-[¹⁵N], Overnight Express™, Sigma-Aldrich®
Catalog # 71759-4
CAS Number:  

Specifications

  • Pk:
    4 SET
  • Seq:
    0167

Specifications

About this item

 :
CAS Number:39466-62-1
Formula:H₄ClN
MDL Number:MFCD00075417