Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
D(+)-Glucose-[6,6-D2]
D-(+)-Glucose-[6,6-D₂] (98% D), Sigma-Aldrich®
D-(+)-Glucose-[6,6-D₂] (98% D), Sigma-Aldrich®
D-(+)-Glucose-[6,6-D₂] (98% D), Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
D-(+)-Glucose-[6,6-D₂] (98% D), Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
18991-62-3
:
Formula:
C₆H₁₀D₂O₆
MDL Number:
MFCD00064367
:
M-Clarity™ quality level = MQ200