Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
D(+)-Glucose-[1-13C]
D-(+)-Glucose-[1-¹³C] (99% ¹³C ), Sigma-Aldrich®
D-(+)-Glucose-[1-¹³C] (99% ¹³C ), Sigma-Aldrich®
D-(+)-Glucose-[1-¹³C] (99% ¹³C ), Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
D-(+)-Glucose-[1-¹³C] (99% ¹³C ), Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
40762-22-9
:
Formula:
C₆H₁₂O₆
MDL Number:
MFCD00037545
:
M-Clarity™ quality level = MQ200