Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
L(-)-Arabitol
L(-)-Arabitol, Sigma-Aldrich®
L(-)-Arabitol, Sigma-Aldrich®
L(-)-Arabitol, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
L(-)-Arabitol, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
7643-75-6
:
Formula:
C₅H₁₂O₅
MDL Number:
MFCD00064290
:
152.15 g/mol
:
M-Clarity™ quality level = MQ300
EINECS:
231-582-6