Order Entry
Germany
ContactUsLinkComponent
L(-)-Arabitol
L(-)-Arabitol
Catalog # MOLE25675490-25G
Supplier:  Molekula
CAS Number:  
L(-)-Arabitol
Catalog # MOLE25675490-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :152.15 g/mol
EINECS:231-582-6
CAS Number:7643-75-6
Formula:C₅H₁₂O₅
MDL Number:MFCD00064290