L(-)-Arabitol 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7643-75-6
:
Formula:C₅H₁₂O₅
MDL Number:MFCD00064290
:152.15 g/mol
Melting Pt:99…103 °C
Storage Temperature:Refrigerator
EINECS:231-582-6
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Melting point:99 to 104 °C
- HPLC:≥98.5 % (ELSD)
- Specific optical rotation:-10° to -13° (20 °C, 589 nm) (c=5, 4% borax)