D-(+)-Arabitol
Catalog # MOLEM78502737
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:152.15 g/mol
EINECS:207-686-2
CAS Number:488-82-4
Formula:C₅H₁₂O₅
MDL Number:MFCD00004709