Order Entry
Germany
ContactUsLinkComponent
D-(+)-Arabitol
D-(+)-Arabitol
Catalog # EHERC10297900
Supplier:  EHRENSTORFER
CAS Number:  
D-(+)-Arabitol
Catalog # EHERC10297900
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :152.15 g/mol
EINECS:207-686-2
CAS Number:488-82-4
Formula:C₅H₁₂O₅
MDL Number:MFCD00004709