Order Entry
Germany
Orders LinkContactUsLinkComponent
Arachidonic acid

Arachidonic acid

Glass bottle for solids, 50 mg
Catalog # SERA13800.01
Marketsource
Supplier:   Serva
Arachidonic acid
Catalog # SERA13800.01
Supplier:  Serva
Marketsource
CAS Number:  506-32-1
Arachidonic acid

Glass bottle for solids, 50 mg

CAS Number:  506-32-1
Price
Inquire For Price
Log in for availability
1 * 50 mg
Product Details & Documents
Synonyms:  (5Z,8Z,11Z,14Z)-Icosa-5,8,11,14-tetraenoic acid
  • CAS Number:506-32-1
  • Formula:C₂₀H₃₂O₂
  • MDL Number:MFCD00004417
  • Molecular Weight:304.5 g/mol
  • EINECS:208-033-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.