Estradiol
Marketsource
Supplier: Serva
Jump to:
About this item
Synonyms: beta-Estradiol, 17β-Estradiol
- CAS Number:50-28-2
- Formula:C₁₈H₂₄O₂
- MDL Number:MFCD00003693
- Molecular Weight:272.39 g/mol
- EINECS:200-023-8
2 Options Available Below
Product Details & Documents
Synonyms: beta-Estradiol, 17β-Estradiol
- CAS Number:50-28-2
- Formula:C₁₈H₂₄O₂
- MDL Number:MFCD00003693
- Molecular Weight:272.39 g/mol
- EINECS:200-023-8
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource



