Order Entry
Germany
ContactUsLinkComponent
Estradiol ≥97%, Calbiochem®, Sigma-Aldrich®
Health
Environment
Estradiol ≥97%, Calbiochem®, Sigma-Aldrich®
Catalog # 3301-1
CAS Number:  
Health
Environment
Estradiol ≥97%, Calbiochem®, Sigma-Aldrich®
Catalog # 3301-1
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :272.39 g/mol
 :M-Clarity™ quality level = MQ100
EINECS:200-023-8
 :GHS08
 :GHS09
Storage Temperature:Refrigerator
CAS Number:50-28-2
Formula:C₁₈H₂₄O₂
MDL Number:MFCD00003693