Order Entry
Germany
ContactUsLinkComponent
(±)-1-Boc-3-aminopyrrolidine, Sigma-Aldrich®
(±)-1-Boc-3-aminopyrrolidine, Sigma-Aldrich®
Catalog # SIAL706582-1G
Supplier:  Merck
CAS Number:  
(±)-1-Boc-3-aminopyrrolidine, Sigma-Aldrich®
Catalog # SIAL706582-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :186.25 g/mol
 :M-Clarity™ quality level = MQ100
CAS Number:186550-13-0
Formula:C₉H₁₈N₂O₂
MDL Number:MFCD01861220