Order Entry
Germany
ContactUsLinkComponent
Dodecacarbonyltriruthenium ≥99%
Dodecacarbonyltriruthenium ≥99%
Catalog # STRE44-1850-1G
CAS Number:  
Dodecacarbonyltriruthenium ≥99%
Catalog # STRE44-1850-1G
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :639.33 g/mol
Melting Pt:150 °C (dec.)
EINECS:239-287-4
UN:3281
ADR:6.1,I
CAS Number:15243-33-1
Formula:C₁₂O₁₂Ru₃
MDL Number:MFCD00011209