Dodecacarbonyltriruthenium ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:15243-33-1
:
Formula:C₁₂O₁₂Ru₃
MDL Number:MFCD00011209
:639.33 g/mol
Melting Pt:∼ 150 °C (dec.)
Storage Temperature:Ambient
EINECS:239-287-4
UN:3466
ADR:6.1,III
:GHS06