Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chloro-4-fluoro-5-nitroanisole
2-Chloro-4-fluoro-5-nitroanisole
2-Chloro-4-fluoro-5-nitroanisole
2-Chloro-4-fluoro-5-nitroanisole
Supplier:
BIONET RESEARCH
Certificates
Expand image
2-Chloro-4-fluoro-5-nitroanisole
Supplier:
BIONET RESEARCH
About this item
CAS Number:
84478-76-2
:
Formula:
C₇H₅ClFNO₃
MDL Number:
MFCD03425810
:
205.5700 g/mol