ContactUsLinkComponent
Safety shoes, slip-on, Bacou ATL Star
Safety shoes, slip-on, Bacou ATL Star
Catalog # 114-4519
Supplier:  HONEYWELL SAFETY
undefined
Safety shoes, slip-on, Bacou ATL Star
Catalog # 114-4519
Supplier:  HONEYWELL SAFETY
Specifications
  • Shoe size:
    41
  • Colour:
    White
  • Pk:
    1 PAIR
Specifications
About this item
These safety shoes, with breathable, water resistant, microfibre upper, are exclusively manufactured for Avantor. This flexible shoe is abrasion and slip resistant and, is therefore, suitable for any working surface.
  • Metal-free, light and flexible, puncture resistant Flexium™ midsole
  • Metal-free athletic AMG toecap (200 J)
  • Anti-static Coolmax® fabric lining with antibacterial silver ions is shock absorbent
  • Dual-density outsole (PU/TPU) maximises shock absorption
  • Elastic strip for a perfect fit around the foot
Colour: White
EN ISO 20345:2007; S3, SRC
Certifications: EN ISO 20345; S3, SRC.