Detergent, heavy duty, floor detergent, opal
TRSTCRY313
- Description:Opal floor detergent for manual cleaning or use in floor scrubbing machines
- Packed:2 x 5 litre plastic containers
- Pk:2
A non foaming, heavy duty, floor detergent to be used manually or through floor scrubbing machines. Opal is excellent at removing organic soils, including fats and oils, and is safe for use on most floor surfaces including vinyl, stone and concrete.
- Heavy duty detergent for use on organic soils
- Suitable for manual use or use in floor scrubbing machines
- Dilution for use: between 1 to 2% v/v
All detergents are supported by comprehensive technical files which provide all the documentation for validation, including residue test methods.
All detergents are supported by comprehensive technical files which provide all the documentation for validation, including residue test methods.
: Supplied with its own certificate of analysis. A detailed technical file (including: product specification, manufacturing process, shelf life, microbiological and analytical data) is available on request.
: Available in 5 litre pack size only.