Tweezers, Azlon®
Supplier: SciLabware
Certificates
About this item
Forceps and tweezers made from PMP or POM.
- High quality, autoclavable
- Available with pointed or rounded ends
- Fine tooth grips for secure handling of samples
Forceps and tweezers made from PMP or POM.