Orders LinkContactUsLinkComponent
Capillary columns, TRACE™ TR-5MS
Capillary columns, TRACE™ TR-5MS
Capillary columns, TRACE™ TR-5MS
Certificates
Capillary columns, TRACE™ TR-5MS
Capillary columns, TRACE™ TR-5MS
7 Options Available Below
Product Details & Documents
TRACE™ TR-5MS GC columns are ideal for hydrocarbons, solvents, pesticides, herbicides, phenols, amines.
  • Non-polar phase, 5% phenyl polysilphenylene-siloxane
  • Low bleed and high stability
  • High signal-to-noise for increased sensitivity
  • High robustness to oxygen and water contamination
Product Family Options
Product Information
  • Column Format
    Phase
    Film thickness
    Ø int.
    Length
    Availability
    Price
  • Capillary
    TR-5MS
    3,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 964,00 €
    964,00 €
    1 * 1 ST
       964,00 €
  • Capillary
    TR-5MS
    0,18 µm
    0,18 mm
    20 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 719,00 €
    719,00 €
    1 * 1 ST
       719,00 €
  • Capillary
    TR-5MS
    0,10 µm
    0,25 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 468,00 €
    468,00 €
    1 * 1 ST
       468,00 €
  • Capillary
    TR-5MS
    0,10 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 850,00 €
    850,00 €
    1 * 1 ST
       850,00 €
  • Capillary
    TR-5MS
    1,00 µm
    0,32 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 499,00 €
    499,00 €
    1 * 1 ST
       499,00 €
  • Capillary
    TR-5MS
    1,00 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 840,00 €
    840,00 €
    1 * 1 ST
       840,00 €
  • Capillary
    TR-5MS
    1,00 µm
    0,25 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 410,00 €
    1 410,00 €
    1 * 1 ST
       1 410,00 €