ContactUsLinkComponent
Ergonomic floor mats, ESD, Ergomat®
Ergonomic floor mats, ESD, Ergomat®
Catalog # ERGOEFB-LIGHTGREY
Supplier:  ERGOMAT
undefined
Ergonomic floor mats, ESD, Ergomat®
Catalog # ERGOEFB-LIGHTGREY
Supplier:  ERGOMAT
Specifications
  • Colour:
    Light grey
  • L×W:
    900×600 mm
  • Seq:
    01
  • Thickness:
    15
  • Pk:
    1
Specifications
About this item
The ESD mats Ergomat® EFB and EFS Complete are made from pure polyurethane and are designed to increase comfort while reducing trip hazards and body pain. These mats are ideal for areas that combine sitting and standing or where static electricity control is a must. The smooth version is ideal for areas where trolleys and carts are used.
  • Choice of smooth or bubbled upper surface
  • Fire retardant, resists moderate oils and liquids
  • Moulded bevelled edges prevent curling, eliminates trip hazards
  • Raw virgin material guarantees durability and ergonomic benefits
  • Solid mat throughout, no hollow bubbles that collapse
  • Anti-static, free from silicone and latex
Suitable for use in class ISO 5 (100) cleanrooms.
Colour: Light grey
Meets EN ISO standards
Certifications: Meets EN ISO standards.