E.Z.N.A.® Plant RNA kit
E.Z.N.A.® Plant RNA kit provides a convenient and rapid method for the isolation of total RNA from a variety of plant samples. This kit provides a homogeniser column for filtration and homogenisation of viscous plant cell lysate by centrifugation in combination with the HiBind® RNA spin column for RNA purification. All the contaminants including polysaccharides and phenolic compounds are effectively removed. Purified RNA can be used for most downstream applications such as RT-PCR, northern blot analysis, differential display, and poly(A)+ RNA selection.
- High quality RNA in 30 minutes
- No organic extractions
- Purified RNA suitable for most applications
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:Total RNA
- Sample type:Plant samples
- Target molecule:RNA
- Sample size:10 - 100 mg
- Description:E.Z.N.A.® Plant RNA Kit