ContactUsLinkComponent
VWR® Chemical Resistant Cleanroom Overalls, Type 4/5/6
VWR® Chemical Resistant Cleanroom Overalls, Type 4/5/6
Supplier:  VWR Collection
Certificates
undefined
VWR® Chemical Resistant Cleanroom Overalls, Type 4/5/6
Supplier:  VWR Collection
Specifications
  • Catalog No:
  • 113-8236
  • 113-8235
  • 113-8232
  • 113-8231
  • 113-8234
  • 113-8233
  • Anti-static:
  • +
  • +
  • +
  • +
  • +
  • +
  • Cleanroom class:
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • Garment application:
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Weight:
  • 60 g/m²
  • 60 g/m²
  • 60 g/m²
  • 60 g/m²
  • 60 g/m²
  • 60 g/m²
Specifications