Orders LinkContactUsLinkComponent
Barrels, wide neck, with lid
Barrels, wide neck, with lid
Supplier:   SEMADENI
Barrels, wide neck, with lid
Supplier:  SEMADENI
Certificates
Barrels, wide neck, with lid
Barrels, wide neck, with lid
3 Options Available Below
Product Details & Documents
Made of HDPE, blue, round (except model 2831 which is rectangular).
  • With handles
  • Good chemical and temperature resistance
  • With UN approval
Heavy duty open top drums with PE lid and galvanised clamping ring. Suitable for storage, disposal and transport of laboratory materials.
The 150 L barrel is specially designed as an ISO container.
Certifications: 30, 60, 120 and 220 L barrels are approved for food use.
Product Family Options
Product Information
  • Capacity
    Ext.Ø
    Ø opening
    Height
    Availability
    Price
  • 30 L
    315 mm
    250 mm
    520 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 84,70 €
    84,70 €
    1 * 1 ST
       84,70 €
  • 60 L
    400 mm
    320 mm
    625 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 80,50 €
    80,50 €
    1 * 1 ST
       80,50 €
  • 120 L
    496 mm
    395 mm
    800 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 163,00 €
    163,00 €
    1 * 1 ST
       163,00 €