Progesterone
Glass bottle for solids, 50 mg
Catalog # PROOBP449
Supplier: LGC Standards PROMOCHEM
Jump to:
Product Details & Documents
- CAS Number:57-83-0
- Formula:C₂₁H₃₀O₂
- MDL Number:MFCD00003658
- EINECS:200-350-6
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:50 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



