Sodium bis(trimethylsilyl)amide 2 M in THF, AcroSeal®, pure
:1070-89-9
:
:C₆H₁₈NNaSi₂
:MFCD00009835
:183.38 g/mol
:–21 °C
:Ambient
:213-983-8
:2924
:3,II
:GHS02
:GHS05
:GHS07
:GHS08
- Appearance:Clear orange to brown Solution
- Titration with HClO4:38 to 42 %