Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Matrix oil, hydrocarbon oil
Matrix oil, hydrocarbon oil, 20 cSt
Matrix oil, hydrocarbon oil, 20 cSt
Matrix oil, hydrocarbon oil, 20 cSt
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Matrix oil, hydrocarbon oil, 20 cSt
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
64742-65-0
:
MDL Number:
MFCD00147539
Storage Temperature:
Ambient
EINECS:
265-169-7
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFA044012
Videos
Thermo Scientific Acroseal 25 Years
Documentation