Order Entry
Germany
ContactUsLinkComponent
Sodium dihydrogen phosphate monohydrate 98.0-102.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Sodium dihydrogen phosphate monohydrate 98.0-102.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Sodium dihydrogen phosphate monohydrate 98.0-102.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    98.0 - 102.0%
  • For Laboratory,Research,or Manufacturing Use:
    4.1 - 4.5
  • Assay (NaH₂PO₄·H₂O):
    98.0 - 102.0 %
  • pH of 5% Solution at 25°C:
    4.1 - 4.5
  • Insoluble Matter:
    ≤0.01 %
  • Chloride (Cl):
    ≤5 ppm
  • ACS - Sulfate (SO₄):
    ≤0.003 %
  • Calcium (Ca):
    ≤0.005 %
  • Potassium (K):
    ≤0.01 %
  • Heavy Metals (as Pb):
    ≤0.001 %
  • Trace Impurities - Iron (Fe):
    ≤0.001 %

Specification Test Results

Related Information

Certificates
Search by Lot Number