Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Nicotinamide

Glass bottle for solids, 100 mg
Catalog # PROOBP460
Irritant
Nicotinamide
Catalog # PROOBP460
CAS Number:  98-92-0
Nicotinamide

Glass bottle for solids, 100 mg

CAS Number:  98-92-0
Price
303,00 €
Log in for availability
1 * 100 mg
Product Details & Documents
Synonyms:  3-(Aminocarbonyl)pyridine, Niacinamide, pyridine-3-carboxamide, Vitamin B3
  • CAS Number:98-92-0
  • Formula:C₆H₆N₂O
  • MDL Number:MFCD00006395
  • Molecular Weight:122.13 g/mol
  • Merck Index:12,06574
  • Boiling Pt:334 °C (1013 hPa)
  • Melting Pt:127…128 °C
  • Density:1.4463 g/cm³ (20 °C)
  • Flash Pt:182 °C
  • Storage Temperature:Ambient
  • EINECS:202-713-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg

Specifications