Order Entry
Germany
ContactUsLinkComponent
Bovine desoxyribonuclease II Typ V (from Spleen)
Bovine desoxyribonuclease II Typ V (from Spleen)
Supplier:  Merck
Certificates
Bovine desoxyribonuclease II Typ V (from Spleen)
Supplier:  Merck

Specifications

  • CAS No.:
    9025-64-3
  • Source:
    Spleen
  • Species:
    Bovine
  • Enzyme name:
    Desoxyribonuclease II, Typ V
  • Enzyme synonyms:
    DNase II

Specifications

Frequently Bought Together