Supelco® PESTANAL® Organic Reference Standard, 2,4,5-TP methyl ester
45692-250MG
: Merck
:
- Pk:250 mg
- Standards:PESTANAL®, analytical standard
- CAS No.:4841-20-7
- Description:2,4,5-TP methyl ester
- Storage conditions:2...30 °C
- Hazard class description:H302 - H319 - H413
- Shipping conditions:STD
- MDL No.:MFCD00055246
- M-Clarity quality level:MQ100
- Synonym(s):Methyl 2-(2,4,5-trichlorophenoxy)propionate
- Molecular weight:283,54
- GHS signal: GHS07
: P273 - P301 + P312 + P330 - P305 + P351 + P338