Herbicide reference standard, 2-(2,4,5-Trichlorophenoxy)propionic acid methyl ester 100 µg/ml in hexane, Dr Ehrenstorfer
Catalog # EHERC13495000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- CAS No.:4841-20-7
- Description:2-(2,4,5-Trichlorophenoxy)propionic acid methyl ester 100 µg/ml in hexane
- Formula:C₁₀H₉Cl₃O₃
- Description:2-(2,4,5-Trichlorophenoxy)propionic acid methyl ester