Herbicide reference standard, 2-(2,4,5-Trichlorophenoxy)propionic acid 10 µg/ml in acetonitrile, Dr Ehrenstorfer
EHERL13490000AL
:
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:93-72-1
- Description:2-(2,4,5-Trichlorophenoxy)propionic acid 10 µg/ml in acetonitrile
- Formula:C₉H₇Cl₃O₃
- Description:2-(2,4,5-Trichlorophenoxy)propionic acid