Order Entry
Germany
ContactUsLinkComponent
4'-Isobutyrylacetophenone
4'-Isobutyrylacetophenone
Catalog # PROOMM0002.09
CAS Number:  
4'-Isobutyrylacetophenone
Catalog # PROOMM0002.09
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
CAS Number:103931-20-0
Formula:C₁₂H₁₄O₂