Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
DepNA (Decanoyl p-nitroaniline)
DepNA (Decanoyl p-nitroaniline)
DepNA (Decanoyl p-nitroaniline)
DepNA (Decanoyl p-nitroaniline)
Supplier:
Cayman Chemical
Certificates
Expand image
DepNA (Decanoyl p-nitroaniline)
Supplier:
Cayman Chemical
About this item
CAS Number:
72298-63-6
:
Formula:
C₁₆H₂₄N₂O₃
MDL Number:
MFCD00033992
:
292.38 g/mol