Sodium bis(trimethylsilyl)amide 0.6 mol/l in toluene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1070-89-9
:
Formula:C₆H₁₈NNaSi₂
MDL Number:MFCD00009835
:183.38 g/mol
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:213-983-8